5-Fluoro-2-formylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (5-Fluoro-2-formylphenyl)boronic acid

A359358-1G A359358-25G A359358-5G A359358-100G A359358-10G
103826-224EA 10 USD
103826-224 103837-232 76840-398 76840-400 76840-402 77033-170 77033-172
5-Fluoro-2-formylphenylboronic acid 98%
5-Fluoro-2-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₃
MW: 167.9300 g/mol
Melting Pt: 137…140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09953731
CAS Number: 1256355-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR