5-Chloro-3-pyridineboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Chloropyridine-3-boronic acid, (5-Chloropyridin-3-yl)boronic acid
A202062-250MG
A202062-25G
A202062-5G
A202062-1G
A202062-100G
103817-352EA
11
USD
103817-352
103847-988
77012-682
77086-082
77086-084
77302-788
77890-223
5-Chloro-3-pyridineboronic acid 98%
5-Chloro-3-pyridineboronic acid
Precaución: for Research Use Only
Formula:
C₅H₅BClNO₂ Melting Pt: 255…260 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06798243 CAS Number: 872041-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...