(5-Chloropyridin-2-yl)methanamine 95%

Supplier: Ambeed
Warning

A679026-1G A679026-5G A679026-100MG A679026-10G A679026-250MG
103840-534EA 331 USD
103840-534 76844-730 76844-732 76879-510 76879-512 76879-514
(5-Chloropyridin-2-yl)methanamine 95%
(5-Chloropyridin-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.58 g/mol
Melting Pt: 34…38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697658
CAS Number: 67938-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR