5-Chloro-2-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2-fluorobenzeneboronic acid, (5-Chloro-2-fluorophenyl)metaboric acid, (5-Chloro-2-fluorophenyl)boronic acid, Dihydroxy(5-chloro-2-fluorophenyl)borane

A134023-100G A134023-10G A134023-1G A134023-25G
103801-956EA 5 USD
103801-956 77095-110 77095-112 77298-680 77298-682
5-Chloro-2-fluorophenylboronic acid 97%
5-Chloro-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BClFO₂
MW: 174.37 g/mol
Boiling Pt: 310.8 °C (760 mm Hg)
Melting Pt: 122…127 °C
Flash Pt: 141.8 °C
Storage Temperature: Ambient
MDL Number: MFCD05664225
CAS Number: 352535-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR