5-Chloro-2,4-difluorophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2,4,-difluorophenylboronic acid, (5-Chloro-2,4-difluorophenyl)boronic acid, 5-Chloro-2,4-difluorobenzeneboronic acid

A115750-100MG A115750-250MG A115750-1G
103798-614EA 57 USD
103798-614 103824-474 76869-996 76869-998 77311-292
5-Chloro-2,4-difluorophenylboronic acid 95%
5-Chloro-2,4-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BClF₂O₂
MW: 192.3600 g/mol
Boiling Pt: 297.1 °C (760 mm Hg)
Flash Pt: 133.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038751
CAS Number: 911645-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR