5-Bromo-2-thiopheneboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-thienylboronic acid, 5-Bromothiophene-2-boronic acid, (5-Bromothiophen-2-yl)boronic acid

A151620-250MG A151620-1G
103804-284EA 8 USD
103804-284 103842-436 76812-210 77318-018
5-Bromo-2-thiopheneboronic acid 96%
5-Bromo-2-thiopheneboronic acid

Precaución: for Research Use Only

Formula: C₄H₄BBrO₂S
MW: 206.85 g/mol
Boiling Pt: 342.3 °C (760 mm Hg)
Melting Pt: 95…100 °C
Flash Pt: 160.8 °C
Storage Temperature: Freezer
MDL Number: MFCD02094028
CAS Number: 162607-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR