(5-Bromopyrimidin-2-yl)methanol 95%

Supplier: Ambeed
Warning

A130207-100MG A130207-10G A130207-250MG A130207-5G A130207-1G
103801-392EA 40 USD
103801-392 76811-140 76811-142 76811-144 77298-396 77313-824
(5-Bromopyrimidin-2-yl)methanol 95%
(5-Bromopyrimidin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16610245
CAS Number: 22433-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR