(5-Bromopyridin-2-yl)methanamine dihydrochloride 97%

Supplier: Ambeed
Warning

A105671-100MG A105671-250MG A105671-1G
103796-924EA 64 USD
103796-924 77008-588 77008-590 77309-526
(5-Bromopyridin-2-yl)methanamine dihydrochloride 97%
(5-Bromopyridin-2-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₉BrCl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD17926556
CAS Number: 1251953-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR