(5-Bromopyridin-2-yl)hydrazine 97%

Supplier: Ambeed
Danger

Synonyms: (5-Bromo-2-pyridyl)hydrazine, 5-Bromo-2-hydrazinopyridine

A105783-100G A105783-10G A105783-25G A105783-1G
103796-942EA 7 USD
103796-942 103822-684 77135-686 77135-688 77135-690 77309-548
(5-Bromopyridin-2-yl)hydrazine 97%
(5-Bromopyridin-2-yl)hydrazine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MW: 188.0252 g/mol
Boiling Pt: 305.7 °C (760 mm Hg)
Melting Pt: 132…137 °C
Flash Pt: 138.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00513424
CAS Number: 77992-44-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR