(5-Bromo-2-methoxyphenyl)methanamine 96%

Supplier: Ambeed
Warning

A204601-100MG A204601-250MG A204601-1G
103817-562EA 20 USD
103817-562 103848-224 76770-278 76770-280 77086-398
(5-Bromo-2-methoxyphenyl)methanamine 96%
(5-Bromo-2-methoxyphenyl)methanamine

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MW: 216.08 g/mol
Boiling Pt: 282.9 °C (760 mm Hg)
Flash Pt: 124.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07786723
CAS Number: 166530-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR