5-Bromo-2-fluorobenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (5-Bromo-2-fluorophenyl)methanol

A396609-1G A396609-5G A396609-100G A396609-250MG
103826-988EA 9 USD
103826-988 103838-028 76841-452 76841-454 77142-834 77142-836
5-Bromo-2-fluorobenzyl alcohol 97%
5-Bromo-2-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 243.8 °C (760 mm Hg)
Melting Pt: 221…226 °C
Flash Pt: 101.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00143425
CAS Number: 99725-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR