5-Bromo-2-chlorobenzyl alcohol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(5-Bromo-2-chlorophenyl)methanol
A227323-10G
A227323-250MG
A227323-1G
A227323-25G
A227323-5G
103819-314EA
6
USD
103819-314
103850-220
76771-036
76771-038
77088-984
77088-986
77088-988
5-Bromo-2-chlorobenzyl alcohol 97%
5-Bromo-2-chlorobenzyl alcohol
Precaución: for Research Use Only
Formula:
C₇H₆BrClO MW: 221.48 g/mol Boiling Pt: 295.8 °C (760 mm Hg) Melting Pt: 92…96 °C Flash Pt: 132.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683548 CAS Number: 149965-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...