3-(Trifluoromethyl)-5-pyridineboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (5-(Trifluoromethyl)pyridin-3-yl)boronic acid, (5-Trifluoromethylpyridin-3-yl)boronic acid

A164306-100MG A164306-250MG A164306-1G
103805-840EA 91 USD
103805-840 103844-100 77028-740 77028-742 77300-704
3-(Trifluoromethyl)-5-pyridineboronic acid 95%
3-(Trifluoromethyl)-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₃NO₂
MW: 190.9156 g/mol
Boiling Pt: 299.9 °C (760 mm Hg)
Melting Pt: 260 °C
Flash Pt: 135.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09952041
CAS Number: 947533-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR