5-Methoxycarbonyl-2-methylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (5-(Methoxycarbonyl)-2-methylphenyl)boronic acid, 5-(Methoxycarbonyl)-2-methylphenylboronic acid

A139961-100MG A139961-250MG A139961-5G A139961-1G
103802-804EA 89 USD
103802-804 76767-724 76767-726 76767-728 77315-912
5-Methoxycarbonyl-2-methylphenylboronic acid 98%
5-Methoxycarbonyl-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MW: 193.992 g/mol
Boiling Pt: 375.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11111429
CAS Number: 876189-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR