5-(Hydroxymethyl)thiophene-3-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (5-(Hydroxymethyl)thiophen-3-yl)boronic acid

A174419-250MG A174419-100MG
103806-942EA 95 USD
103806-942 103845-242 77011-712 77683-403
5-(Hydroxymethyl)thiophene-3-boronic acid 98%
5-(Hydroxymethyl)thiophene-3-boronic acid

Precaución: For research use only.

Formula: C₅H₇BO₃S
MW: 157.983 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27665427
CAS Number: 1268683-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR