(5-(Hydroxymethyl)furan-2-yl)boronic acid 97%

Supplier: Ambeed
Warning

A180047-100MG A180047-250MG
103807-502EA 119 USD
103807-502 103845-856 76824-832 77683-446
(5-(Hydroxymethyl)furan-2-yl)boronic acid 97%
(5-(Hydroxymethyl)furan-2-yl)boronic acid

Precaución: For research use only.

Formula: C₅H₇BO₄
MW: 141.92 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03095371
CAS Number: 1256355-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR