2-(Ethoxycarbonyl)furan-5-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (5-(Ethoxycarbonyl)furan-2-yl)boronic acid

A176996-250MG A176996-5G
103807-196EA 52 USD
103807-196 103845-522 76985-650 77082-896
2-(Ethoxycarbonyl)furan-5-boronic acid 98%
2-(Ethoxycarbonyl)furan-5-boronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₅
MW: 183.9544 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10696632
CAS Number: 1150114-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR