(4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyloxazolidine-5-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4S,5R)-3-Boc-2,2-dimethyl-4-phenyl-1,3-oxazolidine-5-carboxylic acid, (4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyl-1,3-oxazolidine-5-carboxylic acid

A108901-10G A108901-1G
103797-402EA 24 USD
103797-402 103823-192 76995-094 77310-024
(4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyloxazolidine-5-carboxylic acid 95%
(4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyloxazolidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₅
MW: 321.3700 g/mol
Boiling Pt: 459 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09263630
CAS Number: 143527-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR