(S,S)-(-)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) 97%

Supplier: Ambeed
Warning

Synonyms: (S,S)-(-)-2,2'-(Dimethylmethylene)bis(4-tert-butyl-2-oxazoline), (4S,4'S)-2,2'-(Propane-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole), (S,S)-(-)-2,2-Bis(4-tert-butyl-2-oxazolin-2-yl)propane, 2,2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline]

A170834-1G A170834-5G A170834-250MG A170834-25G
103799-420EA 41 USD
103799-420 103806-550 103844-826 77081-948 77081-950 77286-848 77748-130
(S,S)-(-)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) 97%
(S,S)-(-)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline)

Precaución: for Research Use Only

Formula: C₁₇H₃₀N₂O₂
MW: 294.44 g/mol
Melting Pt: 89…91 °C
Flash Pt: 134.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00192243
CAS Number: 131833-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR