(4R,6R)-tert-Butyl-6-(2-aminoethyl)-2,2-dimethyl-1,3-dioxane-4-acetate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-[(4R,6R)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate, 2-[(4R,6R)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetic acid tert-Butyl ester

A100878-100G A100878-10G A100878-25G A100878-5G A100878-500G
103821-360EA 7 USD
103821-360 77233-056 77233-058 77233-060 77296-876 77745-767
(4R,6R)-tert-Butyl-6-(2-aminoethyl)-2,2-dimethyl-1,3-dioxane-4-acetate 97%
(4R,6R)-tert-Butyl-6-(2-aminoethyl)-2,2-dimethyl-1,3-dioxane-4-acetate

Precaución: for Research Use Only

Formula: C₁₄H₂₇NO₄
MW: 273.3700 g/mol
Boiling Pt: 339.8 °C (760 mm Hg)
Flash Pt: 96.8 °C
Storage Temperature: Ambient
MDL Number: MFCD07369252
CAS Number: 125995-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR