(R,R)-(+)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) 97%

Supplier: Ambeed
Warning

Synonyms: (R,R)-(+)-2,2-Bis(4-tert-butyl-2-oxazolin-2-yl)propane, (R,R)-(+)-2,2'-(Dimethylmethylene)bis(4-tert-butyl-2-oxazoline), (4R,4'R)-2,2'-(Propane-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

A467812-250MG A467812-1G A467812-5G A467812-100MG
103827-994EA 227 USD
103827-994 103839-068 77143-640 77307-454 77307-456 77746-956
(R,R)-(+)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) 97%
(R,R)-(+)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline)

Precaución: for Research Use Only

Formula: C₁₇H₃₀N₂O₂
MW: 294.44 g/mol
Boiling Pt: 350.424 °C (760 mm Hg)
Flash Pt: 134.912 °C
Storage Temperature: Ambient
MDL Number: MFCD07368371
CAS Number: 131833-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR