(4-Pyridyl)acetone 98%

Supplier: Ambeed
Warning

A211038-5G A211038-10G A211038-1G A211038-25G A211038-100G
103818-070EA 9 USD
103818-070 103848-764 77087-170 77249-100 77303-168 77303-170 77746-285
(4-Pyridyl)acetone 98%
(4-Pyridyl)acetone

Precaución: for Research Use Only

Formula: C₈H₉NO
Boiling Pt: 232.5 °C (760 mm Hg)
Melting Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00129043
CAS Number: 6304-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR