4-Propylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-n-Propylbenzeneboronic acid, (4-Propylphenyl)boronic acid, 4-n-Propylphenylboronic acid

A463345-100G A463345-10G A463345-1G A463345-25G A463345-5G
103827-950EA 6 USD
103827-950 103839-026 76695-374 76695-376 76842-922 76842-924 76842-926
4-Propylphenylboronic acid 98%
4-Propylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₂
Boiling Pt: 299.1 °C (760 mm Hg)
Melting Pt: 89…97 °C
Flash Pt: 134.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00859261
CAS Number: 134150-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR