(4-Phenylcyclohexyl)methanol 97%

Supplier: Ambeed
Warning

A228177-100MG A228177-250MG
103819-368EA 291 USD
103819-368 103850-276 77139-996 77139-998
(4-Phenylcyclohexyl)methanol 97%
(4-Phenylcyclohexyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₈O
Boiling Pt: 310.7±11.0 °C (760 mm Hg)
Melting Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00610041
CAS Number: 83811-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR