4-Phenoxyphenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4-Phenoxyphenyl)boronic acid, 4-Phenoxybenzeneboronic acid
A380617-100G
A380617-10G
A380617-500G
A380617-1G
A380617-5G
A380617-25G
103826-674EA
5
USD
103826-674
103837-708
76803-714
76803-716
76803-718
76841-028
76841-030
77306-916
4-Phenoxyphenylboronic acid 98%
4-Phenoxyphenylboronic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₁BO₃ Boiling Pt: 377 °C (760 mm Hg) Melting Pt: 141…145 °C Flash Pt: 181.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00093312 CAS Number: 51067-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...