4-Nitrobenzylamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (4-Nitrophenyl)methanamine hydrochloride

A188055-1G A188055-25G A188055-5G A188055-10G A188055-250MG
103816-068EA 16 USD
103816-068 103846-658 77084-380 77084-382 77084-384 77191-858 77191-860
4-Nitrobenzylamine hydrochloride 97%
4-Nitrobenzylamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O₂
MW: 188.62 g/mol
Melting Pt: 265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012863
CAS Number: 18600-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR