4-Nitrophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Nitrobenzeneboronic acid, (4-Nitrophenyl)boronic acid, Dihydroxy(4-nitrophenyl)borane, (4-Nitrophenyl)metaboric acid

A110171-100G A110171-10G A110171-250MG A110171-25G A110171-1G A110171-5G
103797-630EA 7 USD
103797-630 103823-436 77160-692 77160-694 77160-696 77160-698 77310-236 77310-238
4-Nitrophenylboronic acid 97%
4-Nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BNO₄
Boiling Pt: 373.7 °C (760 mm Hg)
Melting Pt: 285…290 °C
Flash Pt: 179.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00161360
CAS Number: 24067-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR