4-(Morpholino)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Morpholinophenyl)boronic acid

A418226-1G A418226-5G A418226-100G A418226-10G A418226-250MG A418226-25G
103827-350EA 14 USD
103827-350 103838-394 76841-994 76841-996 77004-076 77004-078 77004-080 77307-198
4-(Morpholino)phenylboronic acid 98%
4-(Morpholino)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BNO₃
MW: 207.0341 g/mol
Boiling Pt: 425.9 °C (760 mm Hg)
Melting Pt: 126 °C
Flash Pt: 211.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095169
CAS Number: 186498-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR