4-Methyl-3-(hydroxymethyl)morpholine 97%

Supplier: Ambeed
Warning

Synonyms: (4-Methylmorpholin-3-yl)methanol

A139725-100MG A139725-250MG
103802-766EA 130 USD
103802-766 77190-344 77190-346
4-Methyl-3-(hydroxymethyl)morpholine 97%
4-Methyl-3-(hydroxymethyl)morpholine

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.173 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26792530
CAS Number: 1159598-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR