(4-Methylmorpholin-2-yl)methanamine 95%

Supplier: Ambeed
Warning

A155354-100MG A155354-1G
103804-750EA 87 USD
103804-750 103842-946 77273-122 77300-114
(4-Methylmorpholin-2-yl)methanamine 95%
(4-Methylmorpholin-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂O
MW: 130.1882 g/mol
Boiling Pt: 188.7 °C (760 mm Hg)
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03990582
CAS Number: 141814-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR