(4-Methyl-2-(trifluoromethyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A100019-100MG A100019-250MG A100019-1G
103795-200EA 126 USD
103795-200 103820-922 77025-096 77025-098 77282-404
(4-Methyl-2-(trifluoromethyl)phenyl)boronic acid 98%
(4-Methyl-2-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11616521
CAS Number: 1021860-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR