4-Methyl-1H-indazole-5-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Methyl-1H-indazol-5-yl)boronic acid

A150546-250MG A150546-5G
103804-178EA 82 USD
103804-178 103842-314 76973-088 77317-834
4-Methyl-1H-indazole-5-boronic acid 98%
4-Methyl-1H-indazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MW: 175.9800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12028644
CAS Number: 1245816-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR