(4-Methoxypyridin-2-yl)methanamine 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Aminomethyl)-4-methoxypyridine

A100929-100MG A100929-250MG
103795-708EA 86 USD
103795-708 103821-396 76780-424 76780-426
(4-Methoxypyridin-2-yl)methanamine 97%
(4-Methoxypyridin-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.1700 g/mol
Boiling Pt: 242.851 °C (760 mm Hg)
Flash Pt: 100.673 °C
Storage Temperature: Freezer
MDL Number: MFCD11111886
CAS Number: 194658-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR