(4-Methoxybenzyl)hydrazine hydrochloride 97%

Supplier: Ambeed
Warning

A193694-1G A193694-25G A193694-5G A193694-100G A193694-10G A193694-500G
103816-566EA 5 USD
103816-566 103847-180 77085-078 77085-080 77085-082 77139-142 77139-144 77139-146
(4-Methoxybenzyl)hydrazine hydrochloride 97%
(4-Methoxybenzyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃ClN₂O
MW: 188.6546 g/mol
Boiling Pt: 311.9 °C (760 mm Hg)
Melting Pt: 194…195 °C
Flash Pt: 142.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01722406
CAS Number: 2011-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR