4-Methoxy-2-methylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-2-methylbenzeneboronic acid, (4-Methoxy-2-methylphenyl)boronic acid

A180606-5G A180606-10G A180606-1G
103807-546EA 9 USD
103807-546 103845-906 77083-404 77191-668 77301-682
4-Methoxy-2-methylphenylboronic acid 97%
4-Methoxy-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MW: 165.99 g/mol
Boiling Pt: 327 °C (760 mm Hg)
Melting Pt: 169…174 °C
Flash Pt: 151.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02684315
CAS Number: 208399-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR