4-Iodobenzyl alcohol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4-Iodophenyl)methanol
A363852-1G
A363852-25G
A363852-5G
A363852-100G
A363852-10G
A363852-500G
103826-350EA
5
USD
103826-350
103837-368
76840-584
76840-586
76840-588
77106-942
77106-944
77106-946
4-Iodobenzyl alcohol 98%
4-Iodobenzyl alcohol
Precaución: for Research Use Only
Formula:
C₇H₇IO Boiling Pt: 267.1 °C (760 mm Hg) Melting Pt: 72…75 °C Flash Pt: 115.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD01732720 CAS Number: 18282-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...