4-Iodophenylhydrazine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4-Iodophenyl)hydrazine hydrochloride
A311546-1G
A311546-25G
A311546-5G
A311546-100G
A311546-10G
103814-674EA
6
USD
103814-674
103835-866
76838-668
76838-670
76838-672
76977-912
76977-914
4-Iodophenylhydrazine hydrochloride 97%
4-Iodophenylhydrazine hydrochloride
Precaución: For research use only
Formula:
C₆H₇IN₂·ClH Storage Temperature: Refrigerator |
MDL Number:
MFCD00060724 CAS Number: 62830-55-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...