4-Iodobenzophenone 95%

Supplier: Ambeed
Warning

Synonyms: (4-Iodophenyl)(phenyl)methanone, p-Iodobenzophenone

A290841-100MG A290841-250MG A290841-1G A290841-10G
103813-878EA 39 USD
103813-878 103835-020 76723-502 76723-504 77305-584 77668-288
4-Iodobenzophenone 95%
4-Iodobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₉IO
MW: 308.12 g/mol
Boiling Pt: 366.6 °C (760 mm Hg)
Flash Pt: 175.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00270109
CAS Number: 6136-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR