4-Hydroxy-3-methylbenzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-Hydroxy-3-methylphenyl)boronic acid

A155536-100MG A155536-250MG A155536-1G A155536-5G
103804-776EA 70 USD
103804-776 103842-972 76871-828 76871-830 77318-696 77318-698
4-Hydroxy-3-methylbenzeneboronic acid 97%
4-Hydroxy-3-methylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₃
MW: 151.956 g/mol
Boiling Pt: 358 °C (760 mm Hg)
Flash Pt: 170.3 °C
Storage Temperature: Freezer
MDL Number: MFCD09954104
CAS Number: 762263-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR