4-Aminobenzaldehyde, N-BOC protected 97%

Supplier: Ambeed
Warning

A164452-10G A164452-250MG A164452-1G A164452-25G A164452-5G
103805-864EA 22 USD
103805-864 103844-122 77138-270 77138-272 77320-286 77320-288 77320-290
4-Aminobenzaldehyde, N-BOC protected 97%
4-Aminobenzaldehyde, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
Boiling Pt: 297.6 °C (760 mm Hg)
Melting Pt: 138…142 °C
Flash Pt: 133.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06245541
CAS Number: 144072-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR