4-Formyl-2-methoxyphenylboronic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4-Formyl-2-methoxyphenyl)boronic acid
A185411-100MG
A185411-10G
A185411-250MG
A185411-1G
A185411-5G
103815-848EA
18
USD
103815-848
103846-414
76813-716
76813-718
76813-720
77301-968
77301-970
4-Formyl-2-methoxyphenylboronic acid 95%
4-Formyl-2-methoxyphenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₉BO₄ MW: 179.966 g/mol Boiling Pt: 400.7±55.0 °C (760 mm Hg) Melting Pt: 152…155 °C Flash Pt: 196.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11856026 CAS Number: 1028479-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...