4-Fluorophenylhydrazinium chloride 97%

Supplier: Ambeed
Warning

Synonyms: (4-Fluorphenyl)hydrazin HCl, 4-Fluorophenylhydrazine hydrochloride, 1-(4-Fluorophenyl)hydrazine hydrochloride, (4-Fluorophenyl)hydrazine hydrochloride

A337323-25G A337323-100G A337323-10G A337323-500G A337323-5G
103836-718EA 8 USD
103836-718 76839-726 77277-914 77277-916 77277-918 77306-506
4-Fluorophenylhydrazinium chloride 97%
4-Fluorophenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₆H₈ClFN₂
Melting Pt: ≥300 °C
Flash Pt: 90.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00012942
CAS Number: 823-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR