4-Fluoro-4'-methoxybenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: (4-Fluorophenyl)(4-methoxyphenyl)methanone

A128700-100G A128700-10G A128700-500G A128700-25G A128700-5G
76782-190EA 66.84 USD
76782-190 76782-192 76782-194 77313-520 77313-522
4-Fluoro-4'-methoxybenzophenone 98%
4-Fluoro-4'-methoxybenzophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO₂
MW: 230.23 g/mol
Boiling Pt: 351.4 °C (760 mm Hg)
Melting Pt: 90…92 °C
Flash Pt: 160.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00055469
CAS Number: 345-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR