4-Fluoro-2-(trifluoromethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-(trifluoromethyl)benzeneboronic acid, (4-Fluoro-2-(trifluoromethyl)phenyl)boronic acid

A314331-1G A314331-5G A314331-10G
103825-030EA 6 USD
103825-030 103835-950 76838-758 76838-760 77032-660
4-Fluoro-2-(trifluoromethyl)phenylboronic acid 98%
4-Fluoro-2-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₄O₂
MW: 207.92 g/mol
Boiling Pt: 259 °C (760 mm Hg)
Melting Pt: 183…186 °C
Flash Pt: 110.8 °C
Storage Temperature: Ambient
MDL Number: MFCD08062379
CAS Number: 182344-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR