4-Ethynylbenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (4-Ethynylphenyl)methanol

A489164-1G A489164-5G A489164-100MG A489164-10G A489164-250MG
103828-202EA 19 USD
103828-202 103839-274 76843-230 76843-232 77292-040 77292-042 77292-044
4-Ethynylbenzyl alcohol 95%
4-Ethynylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 239.6 °C (760 mm Hg)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD05664205
CAS Number: 10602-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR