(4-Ethynylphenyl)methanamine 95%

Supplier: Ambeed
Warning

A122710-100MG A122710-250MG A122710-1G A122710-5G
103800-258EA 465 USD
103800-258 76870-314 76870-316 77297-884 77297-886
(4-Ethynylphenyl)methanamine 95%
(4-Ethynylphenyl)methanamine

Precaución: for Research Use Only

Formula: C₉H₉N
MW: 131.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10698133
CAS Number: 197844-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR