(4-Ethynylphenyl)(morpholino)methanone 98%

Supplier: Ambeed
Warning

A263467-100MG A263467-250MG A263467-1G
103812-622EA 199 USD
103812-622 103852-510 77014-236 77014-238 77304-874
(4-Ethynylphenyl)(morpholino)methanone 98%
(4-Ethynylphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
MW: 215.25 g/mol
Boiling Pt: 381.2±37.0 °C (760 mm Hg)
Flash Pt: 184.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD23161834
CAS Number: 851895-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR