4-(Ethoxycarbonyl)phenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid
A135283-100G
A135283-10G
A135283-1G
A135283-500G
A135283-25G
A135283-5G
77246-702EA
88
USD
77246-702
77246-704
77246-706
77246-708
77314-924
77314-926
4-(Ethoxycarbonyl)phenylboronic acid 97%
4-(Ethoxycarbonyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₁BO₄ Melting Pt: 135 °C Flash Pt: 168.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179441 CAS Number: 4334-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...