4-Ethoxy-2-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-Ethoxy-2-methoxyphenyl)boronic acid

A556797-1G A556797-100MG A556797-250MG A556797-5G
103828-722EA 27 USD
103828-722 76843-956 77017-720 77017-722 77017-724
4-Ethoxy-2-methoxyphenylboronic acid 97%
4-Ethoxy-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₄
Storage Temperature: Freezer
MDL Number: MFCD04037217
CAS Number: 1207443-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR