2,3-Difluoro-4-ethoxyphenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,3-Difluoro-4-ethoxybenzeneboronic acid, 4-Ethoxy-2,3-difluorobenzeneboronic acid, 4-Ethoxy-2,3-difluorophenylboronic acid, (4-Ethoxy-2,3-difluorophenyl)boronic acid
A168155-1G
A168155-25G
A168155-5G
A168155-10G
103806-256EA
6
USD
103806-256
103844-514
77081-560
77081-562
77081-564
77247-904
2,3-Difluoro-4-ethoxyphenylboronic acid 97%
2,3-Difluoro-4-ethoxyphenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₉BF₂O₃ MW: 201.96 g/mol Boiling Pt: 316 °C (760 mm Hg) Melting Pt: 151…154 °C Flash Pt: 144.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09258743 CAS Number: 212386-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...